Merck
What Is the Cause of Toxicity of Silicone Oil?
Materials (Basel, Switzerland) (2022-01-12)
Ying Chen, Yan Lam Ip, Liangyu Zhou, Pik Yi Li, Yee Mei Chan, Wai Ching Lam, Kenneth Kai Wang Li, David H Steel, Yau Kei Chan
ABSTRACT
To investigate the toxicity of the low-molecular-weight components (LMWCs) in ophthalmic silicone oils (SilOils) on retinal cell lines. The toxicity of six types of LMWCs were studied and compared with conventional SilOil 1000 cSt. In vitro cytotoxic tests of LMWCs, in both liquid and emulsified forms, on three retinal cell lines (MĆ¼ller cells (rMC-1), photoreceptor cells (661W) and retinal pigment epithelial cells (ARPE-19)) were conducted using a transwell cell culturing system. The morphology and viability of cells were assessed by light microscopy and Cell Counting Kit-8 (CCK-8) assay at different time points (6, 24 and 72 h). The ARPE-19 apoptotic pathway was investigated by Mitochondrial Membrane Potential/Annexin V Apoptosis Kit at different time points (6, 24 and 72 h). Apart from dodecamethylpentasiloxane (L5), all liquid LMWCs showed varying degrees of acute cytotoxicity on retinal cell lines within 72 h. Emulsified LMWCs showed comparable cytotoxicity with liquid LMWCs on retinal cell lines. Cyclic LMWCs, octamethylcyclotetrasiloxane (D4) and decamethylcyclopentasiloxane (D5) had significantly higher cytotoxicity when compared with their linear counterparts decamethyltetrasiloxane (L4) and L5 with similar molecular formula. Using ARPE-19 cells as an example, we showed that LMWCs induce the apoptosis of retinal cells. Most LMWCs, in both liquid and emulsified forms, can induce acute cytotoxicity. In addition, cyclic LMWCs are suspected to have higher cytotoxicity than their linear counterparts. Therefore, LMWCs are suspected to be the main cause of the long-term toxicity of ophthalmic SilOil, due to their toxicity and propensity to cause ophthalmic SilOil to emulsify. The amount of LMWCs should be considered as the paramount parameter when referring to the quality of SilOil.
Sigma-Aldrich
Decamethylcyclopentasiloxane, 97%