Merck
  • Gene expression profiling of RIP2-knockdown in HD11 macrophages - elucidation of potential pathways (gene network) when challenged with avian pathogenic E.coli (APEC).
Gene expression profiling of RIP2-knockdown in HD11 macrophages - elucidation of potential pathways (gene network) when challenged with avian pathogenic E.coli (APEC).
BMC genomics (2022-05-04)
Hongyan Sun, Yexin Yang, Yuxuan Cao, Huan Li, Lujiang Qu, Susan J Lamont
ABSTRACT
Receptor interacting serine/threonine kinase 2 (RIP2), ubiquitous in many tissue/cell types, is the key regulator of immune and inflammatory responses for many diseases, including avian pathogenic E. coli (APEC), which causes a wide variety of localized or systemic infections. However, the molecular mechanisms by which RIP2 drives its transcriptional program to affect immune and inflammatory response upon APEC infection remains poorly understood. In this study, RNA-seq and bioinformatics analyses were used to detect gene expression and new direct/indirect RIP2 targets in the treatments of wild type HD11 cells (WT), RIP2 knockdown cells (shRIP2), APEC stimulation cells (APEC), and RIP2 knockdown cells combined with APEC infection (shRIP2 + APEC). The results revealed that a total of 4691 and 2605 differentially expressed genes (DEGs) were screened in shRIP2 + APEC vs. APEC and shRIP2 vs. WT, respectively. Functional annotation analysis showed that apoptosis, MAPK, p53, Toll-like receptor, and Nod-like receptor signaling pathways were involved in APEC-induced RIP2 knockdown HD11 cells. By analyzing the enriched pathway and gene networks, we identified that several DEGs, including HSP90AB1, BID, and CASP9 were targeted by RIP2 upon APEC infection. As a whole, this study can not only provide data support for constructing gene networks of RIP2 knockdown with APEC challenge but also provide new ideas for improving the immune and inflammatory response.