Merck
  • Positive Feedback Regulation of Poly(ADP-ribose) Polymerase 1 and the DNA-PK Catalytic Subunit Affects the Sensitivity of Nasopharyngeal Carcinoma to Etoposide.
Positive Feedback Regulation of Poly(ADP-ribose) Polymerase 1 and the DNA-PK Catalytic Subunit Affects the Sensitivity of Nasopharyngeal Carcinoma to Etoposide.
ACS omega (2022-02-01)
Lingyu Zhang, Yingting Zhuang, Guihui Tu, Ding Li, Yingjuan Fan, Shengnan Ye, Jianhua Xu, Ming Zheng, Ying Wu, Lixian Wu
ABSTRACT
Etoposide (VP-16) is used for the treatment of various cancers, including nasopharyngeal carcinoma (NPC); however, cancers develop resistance to this agent by promoting DNA repair. The DNA-PK (DNA-PKcs) catalytic subunit and poly(ADP-ribose) polymerase 1 (PARP1) mediate acquired resistance and poor survival in NPC cells exposed to DNA damaging agents. DNA repair can alter the sensitivity of NPC cells to DNA damaging agents, and these two enzymes function concomitantly in response to DNA damage in vivo. Therefore, we explored the relationship between DNA-PKcs and PARP1, which may affect NPC cell survival by regulating DNA repair after VP-16 treatment. We performed quantitative real-time polymerase chain reaction, western blotting, and enzyme-linked immunoassays and found that DNA-PKcs knockdown downregulated the PARP1 and PAR expression. Conversely, PARP1 knockdown reduced DNA-PKcs activity, indicating the mutual regulation between DNA-PKcs and PARP1 in VP-16-induced DNA repair. Moreover, a combination treatment with olaparib (a PARP1 inhibitor) and NU7441 (a DNA-PKcs inhibitor) sensitized NPC cells to VP-16 in vitro and in vivo, suggesting that the combined treatment of olaparib, NU7441, and a DNA-damaging agent may be a successful treatment regimen in patients with NPC.
Sigma-Aldrich
Anti-PAR (Ab-1) Mouse mAb (10H), liquid, ≥95% (SDS-PAGE), clone 10H