Merck
  • Fluorescent 6-amino-6-deoxyglycoconjugates for glucose transporter mediated bioimaging.
Fluorescent 6-amino-6-deoxyglycoconjugates for glucose transporter mediated bioimaging.
Biochemical and biophysical research communications (2016-10-26)
Xiangyin Liu, Shengnan Liu, Xinyu Liu, Yunli Shi, Jinna Yang, Zhenhua Huang, Hongxia Zhao, Qingzhi Gao
ABSTRACT
Two novel fluorescent bioprobes, namely, 6N-Gly-Cy3 and 6N-Gly-Cy5, were designed and synthesized for real-time glucose transport imaging as well as potentially useful tracer for galactokinase metabolism. The structure of the bioprobes was fully characterized by 1H NMR, 13C NMR, IR, and HRMS. The fluorescence properties, glucose transporter (GLUT) specificity, and the quenching and safety profiles were studied. The cellular uptake of both bioprobes was competitively diminished by d-glucose, 2-deoxy-d-glucose and GLUT specific inhibitor in a dose-dependent manner in human colon cancer cells (HT29). Comparison study results revealed that the 6N-derived bioprobes are more useful for real-time imaging of cell-based glucose uptake than the structurally similar fluorescent tracer 6-NBDG which was not applicable under physiological conditions. The up to 96 h long-lasting quenching property of 6N-Gly-Cy5 in HT29 suggested the potential applcability of the probe for cell labeling in xenograft transplantation as well as in vivo animal imaging studies.
Sigma-Aldrich
GlucoseCy5, ≥98% (HPLC)