Merck
TiO2-graphene nanocomposites for enhanced osteocalcin induction.
Materials science & engineering. C, Materials for biological applications (2014-03-25)
Kavitha Kandiah, Prabhu Muthusamy, Selvam Mohan, Rajendran Venkatachalam
ABSTRACT
Bone defects and damages are common these days, which increases the usage of biomaterial for humans. To prepare a potential biomaterial, we synthesised a series of titania-graphene nanocomposites (TGS) (2:x (0.25, 0.5, 1.0, 2.0, and 4.0 g)) using in situ sol-gel method. The obtained structural results show that the prepared TGS nanocomposites are an irregular sheet with spherical TiO2 intercalated morphology. The SSA of the nanocomposites ranging from 167.98 to 234.56 m(2) g(-1) with mesoporosity and swelling tendency ranging from 11.55 to 26.13% leads to an enhancement in human cell attachment as well as avoids the migration and agglomeration of the nanoparticles in the body. Further, the biological analysis in simulated body fluid and human cell lines (AGS and MG-63) collectively reveals that the TG2 (2:2) and TG4 (2:4) samples are found to be more favourable materials for biomimic bone action among the prepared TGS nanocomposites.
Product Description
SAFC
Dulbecco′s Modified Eagle′s Medium/Ham′s Nutrient Mixture F12, powder, with 3151 mg/L dextrose, with 2.5 mM L-glutamine, with 55 mg/L sodium pyruvate, without sodium bicarbonate
Pricing and availability is not currently available.