Merck
  • Linked deficiencies in extracellular PP(i) and osteopontin mediate pathologic calcification associated with defective PC-1 and ANK expression.
Linked deficiencies in extracellular PP(i) and osteopontin mediate pathologic calcification associated with defective PC-1 and ANK expression.
Journal of bone and mineral research : the official journal of the American Society for Bone and Mineral Research (2003-06-24)
Kristen Johnson, James Goding, Deborah Van Etten, Adnan Sali, Shou-Ih Hu, David Farley, Hollis Krug, Lovisa Hessle, José Luis Millán, Robert Terkeltaub
ABSTRACT
Osteopontin and PP(i) both suppress hydroxyapatite deposition. Extracellular PP(i) deficiency causes spontaneous hypercalcification, yet unchallenged osteopontin knockout mice have only subtle mineralization abnormalities. We report that extracellular PP(i) deficiency promotes osteopontin deficiency and correction of osteopontin deficiency prevents hypercalcification, suggesting synergistic inhibition of hydroxyapatite deposition. Nucleotide pyrophosphatase phosphodiesterase (NPP) isozymes including PC-1 (NPP1) function partly to generate PP(i), a physiologic calcification inhibitor. PP(i) transport is modulated by the membrane channel protein ANK. Spontaneous articular cartilage calcification, increased vertebral cortical bone formation, and peripheral joint and intervertebral ossific ankylosis are associated with both PC-1 deficiency and expression of truncated ANK in ank/ank mice. To assess how PC-1, ANK, and PP(i) regulate both calcification and cell differentiation, we studied cultured PC-1 -/- and ank/ank mouse calvarial osteoblasts. PC-1 -/- osteoblasts demonstrated approximately 50% depressed NPP activity and markedly lowered extracellular PP(i) associated with hypercalcification. These abnormalities were rescued by transfection of PC-1 but not of the NPP isozyme B10/NPP3. PC-1 -/- and ank/ank cultured osteoblasts demonstrated not only comparable extracellular PP(i) depression and hypercalcification but also marked reduction in expression of osteopontin (OPN), another direct calcification inhibitor. Soluble PC-1 (which corrected extracellular PP(i) and OPN), and OPN itself (> or = 15 pg/ml), corrected hypercalcification by PC-1 -/- and ank/ank osteoblasts. Thus, linked regulatory effects on extracellular PP(i) and OPN expression mediate the ability of PC-1 and ANK to regulate calcification.