Merck
  • Detecting fish parvalbumin with commercial mouse monoclonal anti-frog parvalbumin IgG.
Detecting fish parvalbumin with commercial mouse monoclonal anti-frog parvalbumin IgG.
Journal of agricultural and food chemistry (2006-07-20)
Lingyun Chen, Sue L Hefle, Steve L Taylor, Ines Swoboda, Richard E Goodman
ABSTRACT
Parvalbumin is a calcium-binding muscle protein that is highly conserved across fish species and amphibians. It is the major cross-reactive allergen associated with both fish and frog allergy. We used two-dimensional electrophoretic and immunoblotting techniques to investigate the utility of a commercial monoclonal anti-frog parvalbumin IgG for detecting parvalbumin present in some commonly consumed fish species. The 2D electrophoresis and immunoblots revealed species-specific differences in proteins that appear to represent various numbers of isoforms of parvalbumin in carp (5), catfish (3), cod (1) and tilapia (2). No parvalbumin was detected in yellowfin tuna. Based on minor differences in relative intensities of protein staining and immunodetection, parvalbumin isoforms may have slight differences in the epitope region recognized by the anti-frog parvalbumin antibody. These results suggest that the frog anti-parvalbumin antibody can be used as a valuable tool to detect parvalbumins from the fish tested in this study, except yellowfin tuna.