Merck
Non-apoptotic Fas signaling.
Cytokine & growth factor reviews (2002-12-18)
Harald Wajant, Klaus Pfizenmaier, Peter Scheurich
ABSTRACT
Fas (Apo-1, CD95) and Fas-Ligand (FasL, CD95L) are typical members of the TNF receptor and TNF ligand family, respectively, with a pivotal role in the regulation of apoptotic processes, including activation-induced cell death, T-cell-induced cytotoxicity, immune privilege and tumor surveillance. Impairment of the FasL/Fas system has been implicated in liver failure, autoimmune diseases and immune deficiency. Thus, the FasL/Fas system was mainly appreciated with respect to its death-inducing capabilities. However, there is increasing evidence that activation of Fas can also result in non-apoptotic responses like cell proliferation or NF-kappaB activation. While the apoptotic features of the FasL/Fas system and the pathways involved are comparably well investigated, the pathways that are utilized by Fas to transduce proliferative and activating signals are poorly understood. This review is focused on the non-apoptotic functions of the FasL/Fas system. In particular, the similarities and differences of the molecular mechanisms of apoptotic and non-apoptotic Fas signaling are addressed.
Sigma-Aldrich
Fas Ligand, soluble human, recombinant, expressed in HEK 293 cells, lyophilized powder