Merck
  • Electrospray mass spectrometry for quantitative plasma proteome analysis.
Electrospray mass spectrometry for quantitative plasma proteome analysis.
Methods in molecular biology (Clifton, N.J.) (2009-06-23)
Hong Wang, Sam Hanash
ABSTRACT
Electrospray ionization mass spectrometry (ESI-MS) is an efficient soft ionization procedure for macro biomolecules. However, it is a rather delicate process to produce charged molecules for mass-to-charge ratio (m/z) based measurement. In this chapter, the mechanism of ESI is briefly presented, and the experimental pipeline for quantitative profiling of plasma proteins (prefractionation immunodepletion, protein isotope tagging, 2D-HPLC separation of intact proteins, and LC-MS) is presented as applied by our group in studies of cancer biomarker discovery.
Sigma-Aldrich
Trizma® base, Primary Standard and Buffer, ≥99.9% (titration), crystalline