Merck
Clinical review 125: The insulin receptor and its cellular targets.
The Journal of clinical endocrinology and metabolism (2001-03-10)
Y Kido, J Nakae, D Accili
ABSTRACT
The pleiotropic actions of insulin are mediated by a single receptor tyrosine kinase. Structure/function relationships of the insulin receptor have been conclusively established, and the early steps of insulin signaling are known in some detail. A generally accepted paradigm is that insulin receptors, acting through insulin receptor substrates, stimulate the lipid kinase activity of phosphatidylinositol 3-kinase. The rapid rise in Tris-phosphorylated inositol (PIP(3)) that ensues triggers a cascade of PIP(3)-dependent serine/threonine kinases. Among the latter, Akt (a product of the akt protooncogene) and atypical protein kinase C isoforms are thought to be involved in insulin regulation of glucose transport and oxidation; glycogen, lipid, and protein synthesis; and modulation of gene expression. The presence of multiple insulin-regulated, PIP(3)-dependent kinases is consistent with the possibility that different pathways are required to regulate different biological actions of insulin. Additional work remains to be performed to understand the distal components of insulin signaling. Moreover, there exists substantial evidence for insulin receptor substrate- and/or phosphatidylinositol 3-kinase-independent pathways of insulin action. The ultimate goal of these investigations is to provide clues to the pathogenesis and treatment of the insulin resistant state that is characteristic of type 2 diabetes.