Merck
  • Alterations in the stereochemistry of the kappa-selective opioid agonist U50,488 result in high-affinity sigma ligands.
Alterations in the stereochemistry of the kappa-selective opioid agonist U50,488 result in high-affinity sigma ligands.
Journal of medicinal chemistry (1989-08-01)
B R de Costa, W D Bowen, S B Hellewell, C George, R B Rothman, A A Reid, J M Walker, A E Jacobson, K C Rice
ABSTRACT
The synthesis and in vitro sigma receptor activity of the two diastereomers of U50,488 [(+/-)-2], namely, (1R,2S)-(+)- cis-3,4-dichloro-N-methyl-N-[2-(1-pyrrolidinyl)cyclohexyl]benzeneacet ami de [(+)-1] and (1S,2R)-(-)-cis-3,4-dichloro- N-methyl-N-[2-(1-pyrrolidinyl)cyclohexyl]benzeneacetamide [(-)-1], are described. (+)-1 and (-)-1 were synthesized from (+/-)-trans-N-methyl-2-aminocyclohexanol [(+/-)-3]. Pyridinium chlorochromate (PCC) oxidation of the N-t-Boc-protected derivative of (+/-)-3 afforded (+/-)-2-[N- [(tert-butyloxy)carbonyl]-N-methylamino]cyclohexanone [(+/-)-5]. The sequence of enamine formation with pyrrolidine, catalytic reduction, N-deprotection, and optical resolution afforded (1R,2S)-(-)-cis-2-pyrrolidinyl-N-methylcyclohexylamine [(-)-10] and (1S,2R)-(+)-cis-2-pyrrolidinyl-N-methylcyclohexylamine [(+)-10]. The optical purity (greater than 99.5%) of (-)-10 and (+)-10 was determined by HPLC analysis of the diastereomeric ureas formed by reaction with optically pure (R)-alpha-methylbenzyl isocyanate. The absolute configuration of (-)-10 and (+)-10 was determined by single-crystal X-ray diffractometry of the bis-(R)-mandelate salt. Condensation of optically pure (-)-10 and (+)-10 with 3,4-dichlorophenylacetic acid furnished (+)-1 and (-)-1, respectively. Compounds (+)-1, (-)-1, (-)-2, and (+)-2 were compared for their binding affinities at kappa opioid, sigma, D2-dopamine, and phencyclidine (PCP) receptors in competitive binding assays using [3H]bremazocine ([3H]BREM) or [3H]U69,593, [3H]-(+)-3-(3-hydroxyphenyl)-N-(1-propyl)piperidine [[3H]-(+)-3-PPP], or [3H]-1,3-di(o-tolyl)guanidine ([3H]DTG), [3H]-(-)-sulpiride [[3H]-(-)SULP], and [3H]-1- [1-(2-thienyl)cyclohexyl]piperidine ([3H]TCP), respectively. In the systems examined, (-)-2 exhibited the highest affinity for kappa receptors, with a Ki of 44 +/- 8 nM. However, (-)-2 also showed moderate affinity for sigma receptors, with a Ki of 594 +/- 3 nM [[3H]-(+)-3-PPP]. The (1R,2R)-(+)-enantiomer, (+)-2, had low affinity for both kappa and sigma receptors, exhibiting Ki values of 1298 +/- 49 nM at kappa ([3H]BREM) and 1270 +/- 168 nM at sigma [[3H]-(+)-3-PPP]. In contrast, the chiral cis compounds (+)-1 and (-)-1 showed high affinity for sigma receptors and negligible affinity for kappa opioid receptors in the [3H]BREM assay. Compound (-)-1 exhibited a Ki of 81 +/- 13 nM at sigma receptors [[3H]-(+)-3-PPP] and 250 +/- 8 nM ([3H]DTG).(ABSTRACT TRUNCATED AT 400 WORDS)
Sigma-Aldrich
3,4-Dichlorophenylacetic acid, 98%