Merck
  • Evaluation of the Inclusion of the Green Seaweed Ulva ohnoi as an Ingredient in Feeds for Gilthead Sea Bream (Sparus aurata) and European Sea Bass (Dicentrarchus labrax).
Evaluation of the Inclusion of the Green Seaweed Ulva ohnoi as an Ingredient in Feeds for Gilthead Sea Bream (Sparus aurata) and European Sea Bass (Dicentrarchus labrax).
Animals : an open access journal from MDPI (2021-07-03)
Francisca P Martínez-Antequera, Juan A Martos-Sitcha, Jose M Reyna, Francisco J Moyano
ABSTRACT
This study evaluated the use of Ulva ohnoi as an ingredient in feeds for aquaculture in three different experiments. Experiment 1 was oriented to confirm the negative effect of U. ohnoi on fish digestion. Experiment 2 assessed the effect on growth, feed efficiency, and immune status of juvenile sea bass (Dicentrarchus labrax) fed on diets including U. ohnoi, previously treated or not with carbohydrases used to partially hydrolyze indigestible polysaccharides. Experiment 3 was aimed to evaluate the potential protective effect of U. ohnoi on the oxidative status of sea bream (Sparus aurata) challenged by the consumption of a feed formulated with the oil fraction completely oxidized. Results show a negligible effect of U. ohnoi meal on protein digestion when included in feeds at levels of 10% or less. Moreover, results of growth and feed use evidenced the possibility of using up to 5% inclusion of algal meal in feeds without adverse effects on the zootechnical parameters, while the enzyme pretreatment was ineffective to improve its nutritional use. Finally, the inclusion of U. onhoi in feeds determined both an immunostimulatory effect, evidenced by an increase in skin mucus lysozyme in the two mentioned fish species, and a positive influence on the oxidative metabolism of seabream when fed on a diet including rancid oil.