Merck
  • Evaluation of the cargo contents and potential role of extracellular vesicles in osteoporosis.
Evaluation of the cargo contents and potential role of extracellular vesicles in osteoporosis.
Aging (2021-08-11)
Jifeng Xu, Yu Chen, Dongsheng Yu, Li Zhang, Xiaofan Dou, Gang Wu, Yaping Wang, Shuijun Zhang
ABSTRACT
Osteoporosis is a common aging-related disease diagnosed primarily using bone mineral density (BMD). Extracellular vesicles (EVs) remain unexplored in the context of osteoporosis. Towards this, EVs were isolated from plasma of a discovery cohort with 8 non-osteoporotic and 8 osteoporotic individuals, and nanoparticle tracking analysis (NTA) revealed a significantly higher EV concentration in osteoporotic individuals (P = 0.003). Moreover, EVs concentration showed a linear correlation with bone mineral density (BMD) values (linear correlation coefficient r = 0.9542, deviation from zero, p < 0.001). Results using a mouse model of osteoporosis confirmed that the number of EVs in mice from hindlimb unloading group was significantly higher than that from the age-matched control group (p = 0.015). TaqMan Real-Time PCR demonstrated that miR-335-5p, -320a, -483-5p, and miR-21-5p, were significantly higher expressed in osteoporotic patients compared with non-osteoporotic individuals. Quantitative real-time PCR shown that Wnt1, Wnt5a, Wnt7a, and Wnt9a mRNAs were lower expressed in osteoporosis derived EVs. In vitro functional assay indicated that osteoporosis derived EVs resulted in reduced mineralization in SaOS-2 cells. In conclusion, these results suggest that osteoporosis increased the secretion of EVs which carry higher expression of miRNAs and decreased expression of Wnt signals, further decreased the mineralization capacity in human osteoblasts.