- Endothelial NADPH oxidase 4 protects against angiotensin II-induced cardiac fibrosis and inflammation.
Endothelial NADPH oxidase 4 protects against angiotensin II-induced cardiac fibrosis and inflammation.
Endothelial activation and inflammatory cell infiltration have important roles in the development of cardiac fibrosis induced by renin-angiotensin system activation. NADPH oxidases (Nox proteins) are expressed in endothelial cells (ECs) and alter their function. Previous studies indicated that Nox2 in ECs contributes to angiotensin II (AngII)-induced cardiac fibrosis. However, the effects of EC Nox4 on cardiac fibrosis are unknown. Transgenic (TG) mice overexpressing endothelial-restricted Nox4 were studied alongside wild-type (WT) littermates as controls. At baseline, Nox4 TG mice had significantly enlarged hearts compared with WT, with elongated cardiomyocytes (increased by 18.5%, P < 0.01) and eccentric hypertrophy but well-preserved cardiac function by echocardiography and in vivo pressure-volume analysis. Animals were subjected to a chronic AngII infusion (AngII, 1.1 mg/kg/day) for 14 days. Whereas WT/AngII developed a 2.1-fold increase in interstitial cardiac fibrosis as compared with WT/saline controls (P < 0.01), TG/AngII mice developed significant less fibrosis (1.4-fold increase, P > 0.05), but there were no differences in cardiac hypertrophy or contractile function between the two groups. TG hearts displayed significantly decreased inflammatory cell infiltration with reduced levels of vascular cell adhesion molecule 1 in both the vasculature and myocardium compared with WT after AngII treatment. TG microvascular ECs stimulated with AngII in vitro supported significantly less leukocyte adhesion than WT ECs. A chronic increase in endothelial Nox4 stimulates physiological cardiac hypertrophy and protects against AngII-induced cardiac fibrosis by inhibiting EC activation and the recruitment of inflammatory cells.