Merck
  • Use of esters of N-hydroxysuccinimide in the synthesis of N-acylamino acids.
Use of esters of N-hydroxysuccinimide in the synthesis of N-acylamino acids.
Journal of lipid research (1967-03-01)
Y Lapidot, S Rappoport, Y Wolman
ABSTRACT
Several crystalline N-hydroxysuccinimide esters of short- and long-chain fatty acids have been synthesized. These compounds react with free amino acids to form preferentially N-acylamino acids. The reaction of the N-hydroxysuccinimide esters with hydroxylamine and the behavior of the N-acylamino acids on thin-layer chromatography are described.