Merck
  • Glutamate-induced calcium increase in myotubes depends on up-regulation of a sodium-dependent transporter.
Glutamate-induced calcium increase in myotubes depends on up-regulation of a sodium-dependent transporter.
FEBS letters (2002-09-11)
Claudio Frank, Anna Maria Giammarioli, Loredana Falzano, Carla Fiorentini, Stefano Rufini
ABSTRACT
We report a study on the regulation by 2-chloro adenosine (2CA) of a glutamate (Glu) transporter in myogenic C2C12 cells. Long-term 2CA exposition significantly increased the V(max) of the Glu transporter. Moreover, 2CA-treated cells responded to Glu challenge by a rapid and transient increase in their intracellular calcium level. The above reported effects were totally abolished by treating C2C12 cells with the Na(+)-dependent Glu transporter inhibitors DL-threo-b-hydroxyaspartic acid and L-trans-pyrrolidine-2,4-dicarboxylic acid. We propose that the possible link between the Glu uptake increase and the Glu induction of calcium rise could be the depolarizing currents carried by Na(+) coupled with transporter activity.
Sigma-Aldrich
L-trans-Pyrrolidine-2,4-dicarboxylic acid, ≥98%