Merck
  • Differential effects of resveratrol on the dilator responses of femoral arteries, ex vivo.
Differential effects of resveratrol on the dilator responses of femoral arteries, ex vivo.
Nitric oxide : biology and chemistry (2019-08-02)
Miguel Diaz, Vijay Parikh, Saira Ismail, Raja Maxamed, Emily Tye, Clare Austin, Tristan Dew, Brigitte A Graf, Luc Vanhees, Hans Degens, May Azzawi
ABSTRACT
Resveratrol is a plant-derived phytoalexin with antioxidant, anti-inflammatory and cardio-protective properties and may be a promising therapeutic intervention strategy in cardiovascular disease. Here, we investigated the acute direct effects of trans-resveratrol (RV), on acetylcholine (ACh)-induced and flow-mediated dilation (FMD) of isolated pressurized femoral arteries of young (4-month-old) and old (26-month-old) mice. Vessel exposure to RV enhanced ACh (0.01-1.0 mM)-induced dilation (p < 0.05), but not FMD (@ 5-10 μL⋅min-1) (p < 0.05) in both young and old mice. After RV incubation, acute nitric oxide (NO) production by cultured endothelial cells was increased in response to 0.01 mM ACh, but reduced by flow (5-10 μL⋅min-1; p < 0.05). In isolated femoral arteries from endothelial nitric oxide synthase knockout (eNOS-/-) mice, RV had no overall effect on FMD, but potentiated ACh induced dilation, that was completely abolished by potassium channel blockers, Apamin and Tram 34 (p < 0.01). We demonstrate that the non-metabolised form of RV stimulates ACh-induced dilation via the NO and EDHF pathways, but not FMD by interaction with the cyclo-oxygenase pathway. Our findings have important implications in the use of RV (for both young and aged) under 'normal' non-diseased physiological states.
Sigma-Aldrich
TRAM-34, ≥98% (HPLC), solid
Sigma-Aldrich
Indomethacin, 98.5-100.5% (in accordance with EP)
Sigma-Aldrich
Nitrite/Nitrate Assay Kit, colorimetric, used for detection of nitric oxide metabolites
Sigma-Aldrich
Apamin, from bee venom, ≥95% (HPLC)