Merck
  • BBLN-1 is essential for intermediate filament organization and apical membrane morphology.
BBLN-1 is essential for intermediate filament organization and apical membrane morphology.
Current biology : CB (2021-04-16)
Sanne Remmelzwaal, Florian Geisler, Riccardo Stucchi, Suzanne van der Horst, Milena Pasolli, Jason R Kroll, Olga D Jarosinska, Anna Akhmanova, Christine A Richardson, Maarten Altelaar, Rudolf E Leube, João J Ramalho, Mike Boxem
ABSTRACT
Epithelial tubes are essential components of metazoan organ systems that control the flow of fluids and the exchange of materials between body compartments and the outside environment. The size and shape of the central lumen confer important characteristics to tubular organs and need to be carefully controlled. Here, we identify the small coiled-coil protein BBLN-1 as a regulator of lumen morphology in the C. elegans intestine. Loss of BBLN-1 causes the formation of bubble-shaped invaginations of the apical membrane into the cytoplasm of intestinal cells and abnormal aggregation of the subapical intermediate filament (IF) network. BBLN-1 interacts with IF proteins and localizes to the IF network in an IF-dependent manner. The appearance of invaginations is a result of the abnormal IF aggregation, indicating a direct role for the IF network in maintaining lumen homeostasis. Finally, we identify bublin (BBLN) as the mammalian ortholog of BBLN-1. When expressed in the C. elegans intestine, BBLN recapitulates the localization pattern of BBLN-1 and can compensate for the loss of BBLN-1 in early larvae. In mouse intestinal organoids, BBLN localizes subapically, together with the IF protein keratin 8. Our results therefore may have implications for understanding the role of IFs in regulating epithelial tube morphology in mammals.
Sigma-Aldrich
Anti-C9orf16 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution