Merck
  • Efficacy and Safety of a Proposed Ranibizumab Biosimilar Product vs a Reference Ranibizumab Product for Patients With Neovascular Age-Related Macular Degeneration: A Randomized Clinical Trial.
Efficacy and Safety of a Proposed Ranibizumab Biosimilar Product vs a Reference Ranibizumab Product for Patients With Neovascular Age-Related Macular Degeneration: A Randomized Clinical Trial.
JAMA ophthalmology (2020-11-20)
Se Joon Woo, Miroslav Veith, Jan Hamouz, Jan Ernest, Dominik Zalewski, Jan Studnicka, Attila Vajas, Andras Papp, Vogt Gabor, James Luu, Veronika Matuskova, Young Hee Yoon, Tamás Pregun, Taehyung Kim, Donghoon Shin, Neil M Bressler
ABSTRACT
Neovascular age-related macular degeneration is the leading cause of blindness in individuals 50 years or older. The availability of a ranibizumab biosimilar product (SB11) may facilitate access to an effective alternative to this treatment. To demonstrate equivalence of efficacy, similar safety, and similar immunogenicity of SB11 compared with the reference ranibizumab. This randomized, double-masked, parallel-group phase 3 equivalence study was conducted in 75 centers in 9 countries from March 14, 2018, to December 9, 2019, among 705 participants 50 years or older with neovascular age-related macular degeneration with active subfoveal choroidal neovascularization lesions. Analysis was performed on an intent-to-treat basis. Intravitreous injection of SB11 or ranibizumab, 0.5 mg, every 4 weeks through week 48. Preplanned interim analysis after all participants completed the week 24 assessment of primary efficacy end points at week 8 for change from baseline in best-corrected visual acuity (BCVA) and week 4 for central subfield thickness (CST), with predefined equivalence margins for adjusted treatment differences of -3 letters to 3 letters for BCVA and -36 μm to 36 μm for CST. Baseline and disease characteristics among 705 randomized participants (403 women [57.2%]; mean [SD] age, 74.1 [8.5] years) were comparable between treatment groups (SB11, 351; ranibizumab, 354). Least-squares mean (SE) changes in BCVA from baseline at week 8 were 6.2 (0.5) letters in the SB11 group vs 7.0 (0.5) letters in the ranibizumab group, with an adjusted treatment difference of -0.8 letter (90% CI, -1.8 to 0.2 letters). Least-squares mean (SE) changes in CST from baseline at week 4 were -108 (5) μm in the SB11 group vs -100 (5) μm in the ranibizumab group, with an adjusted treatment difference of -8 μm (95% CI, -19 to 3 μm). Incidences of treatment-emergent adverse events (231 of 350 [66.0%] vs 237 of 354 [66.9%]), including serious treatment-emergent adverse events (44 of 350 [12.6%] vs 44 of 354 [12.4%]) and treatment-emergent adverse events leading to study drug discontinuation (8 of 350 [2.3%] vs 5 of 354 [1.4%]), were similar in the SB11 and ranibizumab groups. Immunogenicity was low, with a cumulative incidence of antidrug antibodies up to week 24 of 3.0% (10 of 330) in the SB11 group and 3.1% (10 of 327) in the ranibizumab group. These findings of equivalent efficacy and similar safety and immunogenicity profiles compared with ranibizumab support the use of SB11 for patients with neovascular age-related macular degeneration. ClinicalTrials.gov Identifier: NCT03150589.
Sigma-Aldrich
3-(1-Pyridinio)-1-propanesulfonate, ≥97.0% (N)