Merck
  • Bioimaging and Biodistribution of the Metal-Ion-Controlled Self-Assembly of PYY3-36 Studied by SPECT/CT.
Bioimaging and Biodistribution of the Metal-Ion-Controlled Self-Assembly of PYY3-36 Studied by SPECT/CT.
Chembiochem : a European journal of chemical biology (2020-07-16)
Panagiota Kalomoiri, Cristina Rodríguez-Rodríguez, Kasper K Sørensen, Marta Bergamo, Katayoun Saatchi, Urs O Häfeli, Knud J Jensen
ABSTRACT
The controlled self-assembly of peptide- and protein-based pharmaceuticals is of central importance for their mode of action and tuning of their properties. Peptide YY3-36 (PYY3-36 ) is a 36-residue peptide hormone that reduces food intake when peripherally administered. Herein, we describe the synthesis of a PYY3-36 analogue functionalized with a metal-ion-binding 2,2'-bipyridine ligand that enables self-assembly through metal complexation. Upon addition of CuII , the bipyridine-modified PYY3-36 peptide binds stoichiometric quantities of metal ions in solution and contributes to the organization of higher-order assemblies. In this study, we aimed to explore the size effect of the self-assembly in vivo by using non-invasive quantitative single-photon emission computed tomography/computed tomography (SPECT/CT) imaging. For this purpose, bipyridine-modified PYY3-36 was radiolabeled with a chelator holding 111 InIII , followed by the addition of CuII to the bipyridine ligand. SPECT/CT imaging and biodistribution studies showed fast renal clearance and accumulation in the kidney cortex. The radiolabeled bipyridyl-PYY3-36 conjugates with and without CuII presented a slightly slower excretion 1 h post injection compared to the unmodified-PYY3-36 , thus demonstrating that higher self-assemblies of the peptide might have an effect on the pharmacokinetics.
Sigma-Aldrich
1-Hydroxy-7-azabenzotriazole solution, ~0.6 M in DMF
Sigma-Aldrich
Borane dimethylamine complex, 97%