- Synthesis of glycyrrhetinic acid derivatives for the treatment of metabolic diseases.
Synthesis of glycyrrhetinic acid derivatives for the treatment of metabolic diseases.
The effect of glycyrrhetinic acid (GA) and GA-derivatives towards 11beta-hydroxysteroid dehydrogenase (11beta-HSD) was investigated. Novel compounds with modifications at positions C-3, C-11 and C-29 of the GA skeleton were prepared. Single crystal X-ray diffraction data of selected substances are reported and discussed.