- Synthesis and biological evaluation of novel pyrazolyl-2,4-thiazolidinediones as anti-inflammatory and neuroprotective agents.
Synthesis and biological evaluation of novel pyrazolyl-2,4-thiazolidinediones as anti-inflammatory and neuroprotective agents.
Bioorganic & medicinal chemistry (2010-02-09)
Amal M Youssef, M Sydney White, Erika B Villanueva, Ibrahim M El-Ashmawy, Andis Klegeris
PMID20138770
ABSTRACT
Novel pyrazolyl-2,4-thiazolidinediones were prepared via the reaction of appropriate pyrazolecarboxaldehydes with 2,4-thiazolidinediones and substituted benzyl-2,4-thiazolidinediones. The resultant compounds were first evaluated for their anti-inflammatory and neuroprotective properties in vitro. The active compounds were further studied in vivo by using the formalin-induced paw edema and the turpentine oil-induced granuloma pouch bioassays. We identified four novel compounds that showed protective effects in vitro at non-toxic concentrations, and were also effective in the animal models of acute and sub-acute inflammation.