Merck
  • Influence of Hepatocellular Carcinoma on Platelet Aggregation in Cirrhosis.
Influence of Hepatocellular Carcinoma on Platelet Aggregation in Cirrhosis.
Cancers (2021-04-04)
Alberto Zanetto, Marco Senzolo, Elena Campello, Cristiana Bulato, Sabrina Gavasso, Sarah Shalaby, Martina Gambato, Alessandro Vitale, Umberto Cillo, Fabio Farinati, Francesco Paolo Russo, Paolo Simioni, Patrizia Burra
ABSTRACT
Hyper-functional platelets are being proposed as a potential therapeutic target in multiple cancers. Whether this can be considered in patients with cirrhosis and hepatocellular carcinoma (HCC) is unknown as their platelet function has not yet been investigated. We evaluated platelet function in cirrhosis patients with HCC. Patients with cirrhosis with and without HCC were prospectively recruited. Platelet aggregation, a marker of platelet function, was assessed by impedance aggregometry with adenosine diphosphate (ADP), arachidonic acid (ASPI), and thrombin (TRAP) stimulation. Plasmatic levels of Von Willebrand factor antigen (VWF) were also determined. One-hundred patients were recruited (50 cirrhotics with and 50 without HCC). Cirrhosis severity by Child class and platelet count were comparable between cirrhotics with and without HCC. Cirrhotics with HCC had higher ADP- (45 vs. 28; p < 0.001), ASPI- (47 vs. 28; p < 0.001), and TRAP- (85 vs. 75; p = 0.01) induced platelet aggregation than cirrhotics without HCC, all indicative of platelet hyper-function. The relatively increased platelet aggregation in patients with HCC was confirmed after adjusting the analysis for platelet count/severity of thrombocytopenia. Levels of VWF were higher in patients with vs. without HCC (348 vs. 267; p = 0.006), particularly in compensated cirrhosis. In patients with cirrhosis, HCC is associated with increased platelet aggregation and higher VWF. The clinical implications of these findings deserve further investigation.
Sigma-Aldrich
Thrombin Receptor Activator Peptide 6, >95% (HPLC)
Sigma-Aldrich
Ser-Phe-Leu-Leu-Arg-Asn-Pro-Asn-Asp-Lys-Tyr-Glu-Pro-Phe, ≥97% (HPLC)