Merck
  • Evaluating rhamnolipid-enhanced washing as a first step in remediation of drill cuttings and petroleum-contaminated soils.
Evaluating rhamnolipid-enhanced washing as a first step in remediation of drill cuttings and petroleum-contaminated soils.
Journal of advanced research (2020-02-20)
Ibukun O Olasanmi, Ronald W Thring
ABSTRACT
Environmental pollution by petroleum hydrocarbons (PHCs) is a severe and widespread problem impacting human health and the environment. To combat this issue, innovative and sustainable treatment methods are required. This research study investigated rhamnolipid-enhanced washing of drill cuttings and petroleum-contaminated soil obtained from northeastern British Columbia in Canada. The efficiency of PHC reduction was analysed and quantified via a Gas Chromatography equipped with a Flame Ionization Detector. Optimum washing conditions for both drill cuttings and petroleum-contaminated soil were temperature of ∼23.5 °C (room temperature), rhamnolipid concentration of 500 mg/L, and a washing time of 30 min. The optimum stirring speed and solution-to-sample ratio for drill cuttings and petroleum-contaminated soil were 100 rpm; 1:1, and 200 rpm; 4:1 respectively. The maximum PHC reduction recorded for total petroleum hydrocarbon and PHC fractions - F2, F3 and F4 were 76.8%, 85.4%, 71.3% and 76.9% respectively for drill cuttings and 58.5%, 48.4%, 63.5% and 59.8% respectively for petroleum-contaminated soil. The results strongly suggest that soil washing is an effective step in the reduction of PHC and can be used as a first step in the treatment of drill cuttings and petroleum-contaminated soils.
Sigma-Aldrich
Rhamnolipids, 90%