Merck
  • Structural and functional comparison of fumarylacetoacetate domain containing protein 1 in human and mouse.
Structural and functional comparison of fumarylacetoacetate domain containing protein 1 in human and mouse.
Bioscience reports (2020-02-19)
Alexander K H Weiss, Andreas Naschberger, Elia Cappuccio, Christina Metzger, Lorenza Mottes, Max Holzknecht, Jill von Velsen, Matthew W Bowler, Bernhard Rupp, Pidder Jansen-Dürr
ABSTRACT
FAH domain containing protein 1 (FAHD1) is a mammalian mitochondrial protein, displaying bifunctionality as acylpyruvate hydrolase (ApH) and oxaloacetate decarboxylase (ODx) activity. We report the crystal structure of mouse FAHD1 and structural mapping of the active site of mouse FAHD1. Despite high structural similarity with human FAHD1, a rabbit monoclonal antibody (RabMab) could be produced that is able to recognize mouse FAHD1, but not the human form, whereas a polyclonal antibody recognized both proteins. Epitope mapping in combination with our deposited crystal structures revealed that the epitope overlaps with a reported SIRT3 deacetylation site in mouse FAHD1.
Sigma-Aldrich
Oxaloacetic acid, ≥97% (HPLC)
Pricing and availability is not currently available.