Merck
  • Application of freeze-dried Yarrowia lipolytica biomass in the synthesis of lipophilic antioxidants.
Application of freeze-dried Yarrowia lipolytica biomass in the synthesis of lipophilic antioxidants.
Biotechnology letters (2020-10-27)
Bartłomiej Zieniuk, Małgorzata Wołoszynowska, Ewa Białecka-Florjańczyk, Agata Fabiszewska
ABSTRACT
The aim of the study was to evaluate the possibility of using Y. lipolytica biomass as a whole-cell catalyst in the synthesis of lipophilic antioxidants, with the example of esterification of five phenolic acids with 1-butanol. Freeze-dried Y. lipolytica biomass was successfully applied as a biocatalyst in the synthesis of esters of phenylpropanoic acid derivatives with 75-98% conversion. However, in the case of phenylacetic acid derivatives, results below 10% were obtained. The biological activity of phenolic acid esters was strongly associated with their chemical structures. Butyl 3-(4-hydroxyphenyl)propanoate showed an IC50 value of 19 mg/ml (95 mM) and TEAC value of 0.427. Among the compounds tested, butyl esters of 3-(4-hydroxyphenyl)propanoic and 4-hydroxyphenylacetic acids exhibited the highest antifungal activity. Lipophilization of phenolic acids achieved by enzymatic esterification creates prospects for using these compounds as food additives with antioxidant properties in lipid-rich food matrices.
Sigma-Aldrich
Lipase acrylic resin, ≥5,000 U/g, recombinant, expressed in Aspergillus niger