Merck
  • Identification of a novel heterozygous truncation mutation in exon 1 of ARHGAP29 in an Indian subject with nonsyndromic cleft lip with cleft palate.
Identification of a novel heterozygous truncation mutation in exon 1 of ARHGAP29 in an Indian subject with nonsyndromic cleft lip with cleft palate.
European journal of dentistry (2014-12-17)
Deepak Chandrasekharan, Arvind Ramanathan
ABSTRACT
Mutations in exon 1 of ARHGAP29, a RhoA specific GTPase have been identified in North American and Filipino subjects with nonsyndromic cleft palate and cleft lip with or without cleft palate. Since the genetic status of ARHGAP29 in Indian subjects with nonsyndromic oral clefts is not known, we designed the present study to investigate the occurrence of the above mutations in them. Total genomic DNA extracted from peripheral blood of 60 subjects with nonsyndromic cleft palate and cleft lip with or without cleft palate, and equal number of control healthy subjects were amplified with primers flanking exon 1 of ARHGAP29 gene and subjected to direct sequencing. Sequencing analysis identified a nonsense mutation in exon 1 of ARHGAP29 that caused substitution of lysine to stop codon at codon position 32 in a subject with nonsyndromic cleft lip with cleft palate. The mutation, however, occurred in heterozygous condition. None of the other subjects carried mutation in this region. The study has thus identified a rare but novel truncation mutation in ARHGAP29 gene for the first time in nonsyndromic oral clefts.