Merck
  • The G-protein-coupled bile acid receptor Gpbar1 (TGR5) suppresses gastric cancer cell proliferation and migration through antagonizing STAT3 signaling pathway.
The G-protein-coupled bile acid receptor Gpbar1 (TGR5) suppresses gastric cancer cell proliferation and migration through antagonizing STAT3 signaling pathway.
Oncotarget (2015-09-30)
Cong Guo, Jia Su, Zhijun Li, Rui Xiao, Jianxun Wen, Yanyan Li, Meng Zhang, Xueting Zhang, Donna Yu, Wendong Huang, Wei-Dong Chen, Yan-Dong Wang
ABSTRACT
Gpbar1 (TGR5), a membrane-bound bile acid receptor, is well known for its roles in regulation of energy homeostasis and glucose metabolism. Here we show that TGR5 is a suppressor of gastric cancer cell proliferation and migration through antagonizing STAT3 signaling pathway. We firstly show that TGR5 activation greatly inhibited proliferation and migration of human gastric cancer cells and strongly induced gastric cancer cell apoptosis. We then found that TGR5 activation antagonized STAT3 signaling pathway through suppressing the phosphorylation of STAT3 and its transcription activity induced by lipopolysaccharide (LPS) or interleukin-6. TGR5 overexpression with ligand treatment inhibited gene expression mediated by STAT3. It suggests that TGR5 antagonizes gastric cancer proliferation and migration at least in part by inhibiting STAT3 signaling. These findings identify TGR5 as a suppressor of gastric cancer cell proliferation and migration that may serve as an attractive therapeutic tool for human gastric cancer.
Sigma-Aldrich
QCM Collagen Cell Invasion Assay, 24-well (8 µm), Colorimetric, The Cell Invasion Assay Kit kuses a 24 well plate with 8 um pores, which is ideal for evaluation of invasive tumor cells.