Merck
  • Identification and characterization of a SARS-CoV-2 specific CD8+ T cell response with immunodominant features.
Identification and characterization of a SARS-CoV-2 specific CD8+ T cell response with immunodominant features.
Nature communications (2021-05-12)
Anastasia Gangaev, Steven L C Ketelaars, Olga I Isaeva, Sanne Patiwael, Anna Dopler, Kelly Hoefakker, Sara De Biasi, Lara Gibellini, Cristina Mussini, Giovanni Guaraldi, Massimo Girardis, Cami M P Talavera Ormeno, Paul J M Hekking, Neubury M Lardy, Mireille Toebes, Robert Balderas, Ton N Schumacher, Huib Ovaa, Andrea Cossarizza, Pia Kvistborg
ABSTRACT
The COVID-19 pandemic caused by SARS-CoV-2 is a continuous challenge worldwide, and there is an urgent need to map the landscape of immunogenic and immunodominant epitopes recognized by CD8+ T cells. Here, we analyze samples from 31 patients with COVID-19 for CD8+ T cell recognition of 500 peptide-HLA class I complexes, restricted by 10 common HLA alleles. We identify 18 CD8+ T cell recognized SARS-CoV-2 epitopes, including an epitope with immunodominant features derived from ORF1ab and restricted by HLA-A*01:01. In-depth characterization of SARS-CoV-2-specific CD8+ T cell responses of patients with acute critical and severe disease reveals high expression of NKG2A, lack of cytokine production and a gene expression profile inhibiting T cell re-activation and migration while sustaining survival. SARS-CoV-2-specific CD8+ T cell responses are detectable up to 5 months after recovery from critical and severe disease, and these responses convert from dysfunctional effector to functional memory CD8+ T cells during convalescence.
Sigma-Aldrich
Biotin, ≥99% (HPLC), lyophilized powder