Merck
  • Lysyl oxidase inhibition via β-aminoproprionitrile hampers human umbilical vein endothelial cell angiogenesis and migration in vitro.
Lysyl oxidase inhibition via β-aminoproprionitrile hampers human umbilical vein endothelial cell angiogenesis and migration in vitro.
Molecular medicine reports (2018-02-03)
Lin Shi, Ning Zhang, Hetao Liu, Lei Zhao, Jing Liu, Juan Wan, Wenyi Wu, Hetian Lei, Rongqing Liu, Mei Han
ABSTRACT
Lysyl oxidase (LOX) is an enzyme that oxidizes lysine residues in collagens and elastin. It stabilizes or remodels the extracellular matrix and basement membrane of blood vessels. Current oncology studies have revealed that LOX is upregulated in invasive cancer cells and bolstered cell movement, and LOX was observed to promote the angiogenesis and migration of endothelial cells. In the present study, angiogenesis and migration were examined in human umbilical vein endothelial cells (HUVECs). Following cell treatment with 0.1-0.4 mM β-aminoproprionitrile (BAPN), a specific inhibitor of LOX, angiogenesis was analyzed with a fibrin gel in vitro angiogenesis assay kit and migration was examined via a Boyden Chamber assay. Angiogenesis-associated gene expression was investigated with a microarray assay and confirmed with reverse transcription-quantitative polymerase chain reaction (RT-qPCR). The results showed that HUVEC angiogenesis substantially increased in the presence of vascular endothelial growth factor (VEGF), basic fibroblast growth factor (bFGF) and phorbol 12-myristate 13-acetate (PMA). In addition, LOX inhibition blocked the angiogenesis stimulated by VEGF bFGF and PMA, and the inhibition of LOX reduced the migration of HUVECs. Furthermore, the microarray and RT-qPCR revealed that BAPN downregulated myeloid progenitor inhibitory factor 1, and western blot analysis demonstrated that BAPN decreased the phosphorylation of MAPK and Akt, suggesting that the specific inhibitor of LOX, BAPN, may serve as an alternative strategy for preventing angiogenesis.