Merck
Boron Uptake Assay in Xenopus laevis Oocytes.
Bio-protocol (2018-03-05)
Sheliang Wang, Namiki Mitani-Ueno, Junpei Takano
ABSTRACT
Boron (B) is essential for plant growth and taken up by plant roots as boric acid. Under B limitation, B uptake and translocation in plants are dependent on the boric acid channels located in the plasma membrane. Xenopus leavis oocyte is a reliable heterologous expression system to characterize transport activities of boric acid channels and related major intrinsic proteins (aquaporins). Here, we outline the protocols for expression of boric acid channels and boric acid uptake assay in Xenopus leavis oocytes.