Merck
  • Berberine analogues as a novel class of the low-density-lipoprotein receptor up-regulators: synthesis, structure-activity relationships, and cholesterol-lowering efficacy.
Berberine analogues as a novel class of the low-density-lipoprotein receptor up-regulators: synthesis, structure-activity relationships, and cholesterol-lowering efficacy.
Journal of medicinal chemistry (2008-12-19)
Ying-Hong Li, Peng Yang, Wei-Jia Kong, Yan-Xiang Wang, Chang-Qin Hu, Zeng-Yan Zuo, Yue-Ming Wang, Hong Gao, Li-Mei Gao, Yan-Chun Feng, Na-Na Du, Ying Liu, Dan-Qing Song, Jian-Dong Jiang
ABSTRACT
Twenty-nine derivatives of berberine (1) or pseudoberberine (2) were designed, semisynthesized, and evaluated for their up-regulatory activity on the low-density-lipoprotein receptor (LDLR) expression. SAR analysis revealed that (i) the methylenedioxy group at the 2- and 3-position is an essential element to keep the activity, (ii) the 7-position quaternary ammonium and planar structure of the compound are activity-required, and (iii) addition of electron-donating groups at the 7- or 13-position reduced the activity. Of the compound 1 analogues, compound 2 exhibited an increased activity on LDLR expression compared to 1. In the hyperlipidemic rats, compound 2 (100 (mg/kg)/day) reduced blood CHO and LDL-c by 42.6% and 49.4%, respectively, more efficient than 1 did (p < 0.01 for both). The results were confirmed in the hyperlipidemic mice. LD(50) of 2 in mice was over 5000 mg/kg (oral). We consider compound 2 a promising cholesterol-lowering drug candidate.
Sigma-Aldrich
Berberine chloride form
Pricing and availability is not currently available.