Merck
  • Neuronal uptake of serum albumin is associated with neuron damage during the development of epilepsy.
Neuronal uptake of serum albumin is associated with neuron damage during the development of epilepsy.
Experimental and therapeutic medicine (2016-07-23)
Zanhua Liu, Jinjie Liu, Suping Wang, Sibo Liu, Yongbo Zhao
ABSTRACT
It is well established that brain blood barrier dysfunction following the onset of seizures may lead to serum albumin extravasation into the brain. However, the effect of albumin extravasation on the development of epilepsy is yet to be fully elucidated. Previous studies have predominantly focused on the effect of albumin absorption by astrocytes; however, the present study investigated the effects of neuronal uptake of albumin in vitro and in kainic acid-induced Sprague-Dawley rat models of temporal lobe epilepsy. In the present study, electroencephalogram recordings were conducted to record seizure onset, Nissl and Evans blue staining were used to detect neuronal damage and albumin extravasation, respectively, and double immunofluorescence was used to explore neuronal absorption of albumin. Cell counting was also conducted in vitro to determine whether albumin contributes to neuronal death. The results of the present study indicated that extravasated serum albumin was absorbed by neurons, and the neurons that had absorbed albumin died and were dissolved 28 days after seizure onset in vivo. Furthermore, significant neuronal death was detected after albumin absorption in vitro in a dose- and time-dependent manner. These results suggested that albumin may be absorbed by neurons following the onset of seizures. Furthermore, the results indicated that neuronal albumin uptake may be associated with neuronal damage and death in epileptic seizures. Therefore, attenuating albumin extravasation following epileptic seizures may reduce brain damage and slow the development of epilepsy.
Sigma-Aldrich
Avidin–FITC from egg white, buffered aqueous solution