Merck
  • Acquisition of selective antitumoral effects of recombinant adeno-associated virus by genetically inserting tumor-targeting peptides into capsid proteins.
Acquisition of selective antitumoral effects of recombinant adeno-associated virus by genetically inserting tumor-targeting peptides into capsid proteins.
Oncology letters (2012-08-01)
Han Saem Lee, Ji Yun Kim, Won Il Lee, Sung Jin Kim, Min Ji Ko, Sunjoo Jeong, Keerang Park, Han Choe, Heuiran Lee
ABSTRACT
Recombinant adeno-associated virus serotype 5 (rAAV5) is considered to be a promising gene transfer vehicle. However, preferential gene delivery to the tumor remains a requirement for cancer treatment. We generated rAAV5 mutants bearing tumor marker-binding peptides and analyzed their properties as viral vectors, as well as their transduction efficiencies and preferential antitumoral potencies. All of the mutants were successfully produced. Transduction analyses showed that rAAV5 mutants harboring tumor-homing peptides, including RGD and TnC, transduced human cancer cells expressing corresponding receptors on their surfaces. RGDS peptides and TnC antibodies significantly suppressed transduction by rAAV5-RGD and rAAV5-TnC. Cytotoxicity was evident upon transfer of HSV-TK to cells by re-targeted rAAV5. These results provide evidence that rAAV5 vectors, genetically armed with tumor-targeting ligands, preferentially infect human cancer cells harboring the corresponding receptors, thereby inducing antitumoral effects. Further optimization of rAAV5 mutant viruses should thus facilitate practical exploitation of these vectors for gene-based cancer treatment.
Sigma-Aldrich
Anti-Integrin αVβ3 Antibody, clone LM609, FITC conjugated, clone LM609, Chemicon®, from mouse
Sigma-Aldrich
Anti-Integrin αVβ5 Antibody, clone P1F6, clone P1F6, Chemicon®, from mouse