Merck
Effects of Selenium in the MAPK Signaling Cascade.
Journal of cardiovascular and thoracic research (2015-10-03)
Nadereh Rashtchizadeh, Pouran Karimi, Parvin Dehgan, Mohamadreza Salimi Movahed
ABSTRACT
This study aimed to discover by which mechanism selenium (Se) suppresses stimulated platelets stimulation in oxidative stress underlying diseases. Human platelets pretreated with Se and stimulated by Cu(2+)-oxidized low density of lipoprotein (OxLDL) or thrombin before assessment of P-selectin and phosphorylated p38 mitogen-activated protein kinase (p-p38MAPK), phosphorylated Jun N-terminal kinase (p- JNK), and phosphorylated extracellular signal-regulated kinases (p-ERK1/2). All variables were measured by solid phase sandwich enzyme-linked immunosorbent assay (ELISA). Se significantly decreased Cu(2+)-OxLDL induced P-selectin expression, as well as p38 and JNK phosphorylation in platelets, but could not significantly reduce ERK1/2 phosphorylation. Se suppresses inflamed platelets. This effect maybe partly mediated by the p38 or c-JNK signaling pathways. These results create possibility of new co-anti-inflammatory insight for Se in atherosclerosis.