Merck
  • Imaging of branched chain amino acid metabolism in tumors with hyperpolarized 13C ketoisocaproate.
Imaging of branched chain amino acid metabolism in tumors with hyperpolarized 13C ketoisocaproate.
International journal of cancer (2009-12-05)
Magnus Karlsson, Pernille R Jensen, René in 't Zandt, Anna Gisselsson, Georg Hansson, Jens Ø Duus, Sebastian Meier, Mathilde H Lerche
ABSTRACT
Powerful analytical tools are vital for characterizing the complex molecular changes underlying oncogenesis and cancer treatment. This is particularly true, if information is to be collected in vivo by noninvasive approaches. In the recent past, hyperpolarized (13)C magnetic resonance (MR) spectroscopy has been employed to quickly collect detailed spectral information on the chemical fate of tracer molecules in different tissues at high sensitivity. Here, we report a preclinical study showing that alpha-ketoisocaproic acid (KIC) can be used to assess molecular signatures of tumors with hyperpolarized MR spectroscopy. KIC is metabolized to leucine by the enzyme branched chain amino acid transferase (BCAT), which is found upregulated in some tumors. BCAT is a putative marker for metastasis and a target of the proto-oncogene c-myc. Very different fluxes through the BCAT-catalyzed reaction can be detected for murine lymphoma (EL4) and rat mammary adenocarcinoma (R3230AC) tumors in vivo. EL4 tumors show a more than 7-fold higher hyperpolarized (13)C leucine signal relative to the surrounding healthy tissue. In R3230AC tumor on the other hand branched chain amino acid metabolism is not enhanced relative to surrounding tissues. The distinct molecular signatures of branched chain amino acid metabolism in EL4 and R3230AC tumors correlate well with ex vivo assays of BCAT activity.
Sigma-Aldrich
Bicinchoninic Acid Kit for Protein Determination, for 200-1000 μg/ml protein