Merck
  • Field-induced SIM behaviour of a Co(II) complex with a 1,1'-diacetylferrocene-derived ligand.
Field-induced SIM behaviour of a Co(II) complex with a 1,1'-diacetylferrocene-derived ligand.
Dalton transactions (Cambridge, England : 2003) (2020-11-04)
Yulia P Tupolova, Igor N Shcherbakov, Leonid D Popov, Roman B Morgunov, Denis V Korchagin, Vladimir E Lebedev, Andrew V Palii, Sergey M Aldoshin
ABSTRACT
Herein, we report the synthesis and magnetic properties of the Co(ii) coordination compound with the 1,1'-bis(1-((pyrid-2-ylmethylene)hydrazono)ethyl)ferrocene (L) ligand, having the general formula [CoLCl2]. The static magnetic data analysis supported by the CASSCF/NEVPT2 calculations revealed the presence of the triaxial magnetic anisotropy with Dexp = +35.2 cm-1 and large rhombicity (E/D = 0.31) in this complex (Dcalc = +34.5 cm-1, E/Dcalc = 0.30). The dynamic magnetic data confirm that the complex shows a slow field-induced (HDC = 1000 Oe) magnetic relaxation behaviour.