- 1-(1-Naphthyl)piperazine as a novel template for 5-HT6 serotonin receptor ligands.
1-(1-Naphthyl)piperazine as a novel template for 5-HT6 serotonin receptor ligands.
Bioorganic & medicinal chemistry letters (2005-03-05)
Mase Lee, Jagadeesh B Rangisetty, Manik R Pullagurla, Małgorzata Dukat, Vince Setola, Bryan L Roth, Richard A Glennon
PMID15745826
ABSTRACT
4-Sulfonyl analogs of 1-(1-naphthyl)piperazine bind at human 5-HT6 receptors and represent a novel class of human 5-HT6 receptor ligands.