Merck
  • Investigations on Metabolic Changes in Beagle Dogs Fed Probiotic Queso Blanco Cheese and Identification of Candidate Probiotic Fecal Biomarkers Using Metabolomics Approaches.
Investigations on Metabolic Changes in Beagle Dogs Fed Probiotic Queso Blanco Cheese and Identification of Candidate Probiotic Fecal Biomarkers Using Metabolomics Approaches.
Metabolites (2020-07-30)
Ye Jin Kim, Ho-Eun Park, Wan-Kyu Lee, Jun-Sang Ham, Sang Un Park, Jae Geun Kim, Kyung-Hoan Im, Jae Kwang Kim
ABSTRACT
Intake of probiotic cheese improves the intestinal health of humans and animals. However, metabolic changes in the intestines of dogs in response to the ingestion of probiotic cheese have not been evaluated. Thus, we aimed to determine the metabolic changes in healthy beagle dogs fed queso blanco cheese with added Lactobacillus reuteri KACC 92293 and Bifidobacterium longum KACC 91563 (QCLB) and to identify potential fecal biomarkers to distinguish the metabolic changes based on intake of probiotic cheese through metabolomics approaches. The dogs were randomly divided into three groups and fed a regular diet without any cheese (control), a diet with queso blanco cheese (QC), or one with QCLB for eight weeks. The concentrations of acetic, propionic, and 4-aminobutyric acids were increased in the QCLB group compared to those in the control group. Additionally, higher levels of propionic acid and lower levels of xylose were found in the QCLB group compared to those in the QC group. This is the first report on the identification of metabolic changes in beagle dogs fed queso blanco cheese with added L. reuteri KACC 92293 and B. longum KACC 91563. We also found that metabolomics approaches can be useful for identifying potential fecal markers in dogs fed probiotic cheese.
Product Description
Supelco
F.A.M.E. Mix, C8 - C24, certified reference material, ampule of 100 mg (Neat)
Sigma-Aldrich
Poly(dimethylsiloxane), viscosity 1.0 cSt (25 °C)
Supelco
SPME Fiber Holder, for use with manual sampling