Merck
  • Atorvastatin Combined with Low-Dose Dexamethasone Treatment Protects Endothelial Function Impaired by Chronic Subdural Hematoma via the Transcription Factor KLF-2.
Atorvastatin Combined with Low-Dose Dexamethasone Treatment Protects Endothelial Function Impaired by Chronic Subdural Hematoma via the Transcription Factor KLF-2.
Drug design, development and therapy (2020-08-28)
Yueshan Fan, Dong Wang, Chenxu Rao, Ying Li, Hongtao Rong, Zengguang Wang, Jianning Zhang
ABSTRACT
Our previous study showed that the combination therapy with atorvastatin and low-dose dexamethasone protected endothelial cell function in chronic subdural hematoma (CSDH) injury. In this study, we aimed to investigate the mechanism underlying the effects of this combination therapy on CSDH-induced cell dysfunction. Monocytes and endothelial cells were cocultured with CSDH patient hematoma samples to mimic the pathological microenvironment of CSDH. Monocytes (THP-1 cells) and endothelial cells (hCMEC/D3 cells) were cocultured in a transwell system for 24 h before stimulation with hematoma samples diluted in endothelial cell medium (ECM) at a 1:1 ratio. Tight junction markers were detected by Western blotting, PCR and immunofluorescence. hCMEC/D3 cells were collected for Western blot and PCR analyses to detect changes in the expression levels of vascular cell adhesion molecule (VCAM-1), intercellular adhesion molecule (ICAM-1), and Kruppel-like factor 2 (KLF-2). The IL-6, IL-10 and VEGF levels in the supernatant were measured by enzyme-linked immunosorbent assay (ELISA). KLF-2 expression in endothelial cells was decreased after stimulation with CSDH patient hematoma samples, but combination therapy with atorvastatin and low-dose dexamethasone reversed this trend. KLF-2 protected injured cells by increasing the expression of VE-cadherin and ZO-1; attenuating the expression of VCAM-1, ICAM-1, IL-6 and VEGF; and enhancing the expression of IL-10, all of which play pivotal roles in endothelial inflammation. Moreover, the effect of combination therapy with atorvastatin and low-dose dexamethasone was obviously reduced in endothelial cells with KLF-2 knockdown compared with normal cells. Coculture with hematoma samples decreased KLF-2 expression in human cerebral endothelial cells. Combination therapy with atorvastatin and low-dose dexamethasone counteracted hematoma-induced KLF-2 suppression in human cerebral endothelial cells to attenuate robust endothelial inflammation and permeability. KLF-2 plays an important role in drug therapy for CSDH and may become the key factor in treatment and prognosis.
Sigma-Aldrich
Dexamethasone, ≥98% (HPLC), powder
Sigma-Aldrich
Collagen Type I, rat tail