- Design of an Interpenetrating Polymeric Network Hydrogel Made of Calcium-Alginate from a Thermos-Sensitive Pluronic Template as a Thermal-Ionic Reversible Wound Dressing.
Design of an Interpenetrating Polymeric Network Hydrogel Made of Calcium-Alginate from a Thermos-Sensitive Pluronic Template as a Thermal-Ionic Reversible Wound Dressing.
Polymer-based hydrogels demonstrate superior performance when used as wound dressing. An ideal dressing should possess an active healing function, absorb wound exudates, and provide a moist interface on the wound for rapid injury repair and the prevention of pain and injury during replacement of the dressing. Thus, the aim of this study was to develop a novel, reversible, smart, interpenetrating polymeric network (IPN) by utilizing the thermosensitive network of pluronic F127 (PF127) as a template to regulate the conformation of calcium-ion-crosslinked alginate. We found that the IPN hydrogels formed soft and elastic thermosensitive networks, retaining their form even after absorbing a large amount of wound exudate. The exterior of the hydrogels was made up of a rigid calcium alginate network that supported the entire hydrogel, promoting the stability of the vascular endothelial growth factor (VEGF) payload and controlling its release when the hydrogel was applied topically to wounds. Raman spectroscopy confirmed the layered structure of the hydrogel, which was found to easily disintegrate even after moderate rinsing of the wound with cold phosphate-buffered saline. Taken together, these results show that the IPN hydrogel developed in this study could be a promising delivery platform for growth factors to accelerate wound healing.