Merck
  • Residue Behavior and Risk Assessment of Rimsulfuron and Quizalofop-P-ethyl in Potato Under Field Conditions.
Residue Behavior and Risk Assessment of Rimsulfuron and Quizalofop-P-ethyl in Potato Under Field Conditions.
Bulletin of environmental contamination and toxicology (2020-09-27)
Shuo Wang, Zhiguang Hou, Shuang Liang, Zhongbin Lu
ABSTRACT
A method for simultaneous quantitation of rimsulfuron, quizalofop-P-ethyl and quizalofop-P in potato plant, soil and potato tuber samples was established. The mean recoveries of rimsulfuron, quizalofop-P-ethyl and quizalofop-P in different matrices spiked with them were 81.4%-101.1%, 76.1%-99.0% and 77.4%-106.4% with relative standard deviations (RSDs) of 2.7%-13.3%, 0.9%-5.5%, 1.7%-11.3%, respectively. The open-field trials in China were conducted in potato cultivation system of Changchun and Jinan. The results indicated that the half-lives of rimsulfuron and quizalofop-P-ethyl were 0.04-13.1 days. The residues of quizalofop-P during the harvest time in Jinan soil were < 0.01-0.044 mg kg-1, while there was no residue of target herbicides detected in all other samples. The risk assessment results demonstrated that the risk quotients (RQs) of rimsulfuron and quizalofop-P-ethyl were 7.857 × 10-5 and 8.730 × 10-3, respectively, which exhibited an acceptable dietary risk to Chinese consumers.
Product Description
Supelco
Rimsulfuron, PESTANAL®, analytical standard
Supelco
Quizalofop-P-ethyl, PESTANAL®, analytical standard