- Proline-rich protein 11 overexpression is associated with a more aggressive phenotype and poor overall survival in ovarian cancer patients.
Proline-rich protein 11 overexpression is associated with a more aggressive phenotype and poor overall survival in ovarian cancer patients.
The proline-rich protein 11 (PRR11) is a newly identified oncogene associated with a poor prognosis in several human cancers. Nonetheless, research on its role in ovarian cancer (OC) remains largely understudied. Therefore, this study aims to evaluate the expression levels of PRR11 protein and its role in human ovarian cancer. Immunohistochemistry analysis was used to evaluate the expression levels of PRR11 protein in human samples obtained from 49 patients diagnosed with OC and subjected to curative surgery in the First Affiliated Hospital of Wenzhou Medical University between 2007 and 2015. In total, 57.1% of the primary OC tumor tissue evaluated demonstrated overexpression of PRR11. Meanwhile, the survival analysis showed that the overall survival (OS) of patients presenting overexpression of PRR11 was significantly lower than the OS of the patients with negative PRR11. In subsequent experiments, it was found that silencing the expression of PRR11 expression inhibited the proliferation of tumor cells and the migration of cells in vitro. Further, cells subjected to PRR11 knockdown exhibited a decrease in tumor growth in vivo. The downregulation of PRR11 was coupled with a decrease in N-cadherin and downregulation in the expression of early growth response protein 1 (EGR1). The findings suggest that PRR11 might be considered as a potential target for prognostic assessment and gene therapy strategies for patients diagnosed with OC.