Merck
  • HSV-1/TLR9-Mediated IFNβ and TNFα Induction Is Mal-Dependent in Macrophages.
HSV-1/TLR9-Mediated IFNβ and TNFα Induction Is Mal-Dependent in Macrophages.
Journal of innate immunity (2019-12-19)
Joanna Zyzak, Małgorzata Mitkiewicz, Ewa Leszczyńska, Patryk Reniewicz, Paul N Moynagh, Jakub Siednienko
ABSTRACT
Innate immune response is a universal mechanism against invading pathogens. Toll-like receptors (TLRs), being part of a first line of defense, are responsible for detecting a variety of microorganisms. Among them TLR9, which is localized in endosomes, acts as a sensor for unmethylated CpG motifs present in bacteria, DNA viruses (e.g., HSV-1), or fungi. TLRs differ from one another by the use of accessory proteins. MyD88 adapter-like (Mal) adapter molecule is considered a positive regulator of TLR2- and TLR4-dependent pathways. It has been reported that this adapter may also negatively control signal transduction induced by TLR3 anchored in the endosome membrane. So far, the role of Mal adapter protein in the TLR9 signaling pathways has not been clarified. We show for the first time that Mal is engaged in TLR9-de-pendent expression of genes encoding IFNβ and TNFα in HSV-1-infected or CpG-C-treated macrophages and requires a noncanonical NF-κB pathway. Moreover, using inhibitor of ERK1/2 we confirmed involvement of these kinases in TLR9-dependent induction of IFNβ and TNFα. Our study points to a new role of Mal in TLR9 signaling through a hitherto unknown mechanism whereby lack of Mal specifically impairs ERK1/2-mediated induction of noncanonical NF-κB pathway and concomitant IFNβ and TNFα production.
Sigma-Aldrich
MISSION® esiRNA, targeting human TIRAP