Merck
  • Angelica sinensis extract protects against ischemia-reperfusion injury in the hippocampus by activating p38 MAPK-mediated p90RSK/p-Bad and p90RSK/CREB/BDNF signaling after transient global cerebral ischemia in rats.
Angelica sinensis extract protects against ischemia-reperfusion injury in the hippocampus by activating p38 MAPK-mediated p90RSK/p-Bad and p90RSK/CREB/BDNF signaling after transient global cerebral ischemia in rats.
Journal of ethnopharmacology (2020-01-29)
Chin-Yi Cheng, Shung-Te Kao, Yu-Chen Lee
ABSTRACT
Angelica sinensis (Oliv.) Diels, commonly known as Dang Gui (DG), is one of the most popular traditional Chinese herbal medicines for the treatment of stroke. However, the effects of DG on transient global cerebral ischemia (GCI) and its precise mechanisms remain unclear. This study aimed to investigate the effects of the DG extract on ischemia-reperfusion (I/R) injury in the hippocampus 7 d after transient GCI and to identify the potential mitogen-activated protein kinase (MAPK)-related signaling pathway in the hippocampus involved in the effects. Rats were intragastrically administered DG at doses of 0.25 g/kg (DG-0.25g), 0.5 g/kg (DG-0.5g), or 1 g/kg (DG-1g) 1, 3, and 5 d after GCI. DG-0.5g and DG-1g treatments effectively promoted hippocampal cornu ammonis 1 (CA1) neuronal survival. DG-0.5g and DG-1g treatments markedly increased phospho-p38 MAPK (p-p38 MAPK), phospho-90-kDa ribosomal S6 kinase (p-p90RSK), cytosolic and mitochondrial phospho-Bad (p-Bad), phospho-cAMP response element-binding protein (p-CREB), brain-derived neurotrophic factor (BDNF), and p-CREB/BDNF expression; decreased 4-hydroxy-2-nonenal, cytochrome c (Cytc), and cleaved caspase-3 expression, and inhibited apoptosis in the hippocampal CA1 region. Pretreatment with a specific inhibitor of p38 MAPK, SB203580, completely blocked the effects of DG-1g on the expression of the aforementioned proteins. DG-0.5g and DG-1g treatments exerted neuroprotective effects on I/R injury by activating p38 MAPK-mediated p90RSK/p-Bad-induced anti-apoptotic-Cytc/caspase-3-related and p90RSK/CREB/BDNF survival signaling in the hippocampus 7 d after transient GCI.