Merck
A human monoclonal antibody blocking SARS-CoV-2 infection.
Nature communications (2020-05-06)
Chunyan Wang, Wentao Li, Dubravka Drabek, Nisreen M A Okba, Rien van Haperen, Albert D M E Osterhaus, Frank J M van Kuppeveld, Bart L Haagmans, Frank Grosveld, Berend-Jan Bosch
ABSTRACT
The emergence of the novel human coronavirus SARS-CoV-2 in Wuhan, China has caused a worldwide epidemic of respiratory disease (COVID-19). Vaccines and targeted therapeutics for treatment of this disease are currently lacking. Here we report a human monoclonal antibody that neutralizes SARS-CoV-2 (and SARS-CoV) in cell culture. This cross-neutralizing antibody targets a communal epitope on these viruses and may offer potential for prevention and treatment of COVID-19.
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Cell Dissociation Solution Non-enzymatic 1x, Prepared in phosphate buffered saline without calcium and magnesium, sterile-filtered, BioReagent, suitable for cell culture
Millipore
Protein G Agarose, Fast Flow, Protein G Agarose, Fast Flow suitable for medium and low pressure chromatography of IgG from mouse, sheep, and rabbit, and for immunoprecipitations.