Merck
  • Soluble Receptor for Glycation End-products Concentration Increases Following the Treatment of Severe Diabetic Ketoacidosis
Soluble Receptor for Glycation End-products Concentration Increases Following the Treatment of Severe Diabetic Ketoacidosis
Journal of clinical research in pediatric endocrinology (2019-09-14)
William H. Hoffman, Takaki Ishikawa, James Blum, Naoto Tani, Tomoya Ikeda, Carol M. Artlett
ABSTRACT
To determine the time relationships of soluble receptor for glycation end-products (sRAGE), [a decoy of the advanced glycation end-products (AGE)-RAGE axis] and D-lactate, (a metabolite of methylglyoxal) in the inflammatory response to diabetic ketoacidosis (DKA). Sixteen children and adolescents with type 1 diabetes (T1D) had blood samples obtained, 6-12 hours into treatment, at three weeks and three months post start of treatment. sRAGE and D-lactate concentrations at three months were considered baseline. Expression of RAGE was investigated in the myocardium of a newly diagnosed and untreated young person with fatal T1D/DKA. sRAGE 6-12 hours after the start of treatment was 39% lower than the values at two weeks (p=0.0036) and at three months (p=0.0023) post treatment. D-lactate was higher during treatment than at three weeks (p=0.04) and at three months (p=0.035). sRAGE concentration was decreased during treatment, compared to concentrations at two weeks and three months after treatment. The increased D-lactate during treatment was in keeping with the known increase in dicarbonyls at this time. The finding of RAGE expression in a young myocardium prior to DKA treatment suggested cardiovascular inflammation pre-treatment and at a young age.