Merck
  • Non-Fullerene Small Molecule Electron-Transporting Materials for Efficient p-i-n Perovskite Solar Cells.
Non-Fullerene Small Molecule Electron-Transporting Materials for Efficient p-i-n Perovskite Solar Cells.
Nanomaterials (Basel, Switzerland) (2020-06-04)
Da-Seul Choi, Sung-Nam Kwon, Seok-In Na
ABSTRACT
PC61BM is commonly used in perovskite solar cells (PSC) as the electron transport material (ETM). However, PC61BM film has various disadvantages, such as its low coverage or the many pinholes that appear due to its aggregation behavior. These faults may lead to undesirable direct contact between the metal cathode and perovskite film, which could result in charge recombination at the perovskite/metal interface. In order to overcome this problem, three alternative non-fullerene electron materials were applied to inverted PSCs; they were evaluated on suitability as electron transport layers. The roles and effects of these non-fullerene ETMs on device performance were studied using photoluminescence (PL) measurements, field emission scanning electron microscopy (FE-SEM), atomic force microscopy (AFM), internal resistance in PSC measurements, and conductive atomic force microscopy (C-AFM). It was found that one of the tested materials, IT-4f, showed excellent electron extraction ability and was associated with reduced recombination. The PSC with IT-4f as the ETM produced better cell-performance; it had an average PCE of 11.21%, which makes it better than the ITIC and COi8DFIC-based devices. Finally, IT-4f was compared with PC61BM; it was found that the two materials have quite comparable efficiency and stability levels.
Sigma-Aldrich
DCB, ≥98% (HPLC), solid
Sigma-Aldrich
Methylammonium bromide, ≥99%, anhydrous