Merck
  • Microbial community dynamics in mesophilic and thermophilic batch reactors under methanogenic, phenyl acid-forming conditions.
Microbial community dynamics in mesophilic and thermophilic batch reactors under methanogenic, phenyl acid-forming conditions.
Biotechnology for biofuels (2020-05-12)
Eva Maria Prem, Blaz Stres, Paul Illmer, Andreas Otto Wagner
ABSTRACT
Proteinaceous wastes exhibit high theoretical methane yields and their residues are considered valuable fertilisers. The routine anaerobic degradation of proteins often raises problems like high aromatic compound concentrations caused by the entry of aromatic amino acids into the system. A profound investigation of the consequences of aromatic compound exposure on various microorganisms, which cascade-like and interdependently degrade complex molecules to biogas, is still pending. In mesophilic samples, methane was predominantly produced via acetoclastic methanogenesis. The highest positive correlation was observed between phenylacetate (PAA) and Psychrobacter spp. and between phenylpropionate (PPA) and Haloimpatiens spp. Moreover, Syntrophus spp. negatively correlated with PAA (Spearman's rank correlations coefficient (rs) = - 0.46, p < 0.05) and PPA concentrations (rs = - 0.44, p < 0.05) and was also associated with anaerobic benzene ring cleavage. In thermophilic samples, acetate was predominantly oxidised by Tepidanaerobacter spp. or Syntrophaceticus spp. in syntrophic association with a hydrogenotrophic methanogen. The genera Sedimentibacter and Syntrophaceticus correlated positively with both PAA and PPA concentrations. Moreover, Sedimentibacter spp., Tepidanaerobacter spp., Acetomicrobium spp., and Sporanaerobacter spp. were significant LEfSe (linear discriminant analysis effect size) biomarkers for high meso- as well as thermophilic phenyl acid concentrations. Direct negative effects of phenyl acids on methanogenic properties could not be proven. Anaerobic phenyl acid formation is not restricted to specific microbial taxa, but rather done by various meso- and thermophilic bacteria. The cleavage of the highly inert benzene ring is possible in methanogenic batch reactors-at least in mesophilic fermentation processes. The results indicated that phenyl acids rather affect microorganisms engaged in preceding degradation steps than the ones involved in methanogenesis.
Sigma-Aldrich
Phenyl propionate, AldrichCPR