Merck
  • QCAL--a novel standard for assessing instrument conditions for proteome analysis.
QCAL--a novel standard for assessing instrument conditions for proteome analysis.
Journal of the American Society for Mass Spectrometry (2008-07-05)
Claire E Eyers, Deborah M Simpson, Stephen C C Wong, Robert J Beynon, Simon J Gaskell
ABSTRACT
If proteome datasets are to be collated, shared, and merged for higher level proteome analyses, there is a need for generally accepted strategies and reagents for optimization and standardization of instrument performance. At present, there is no single protein or peptide standard set that is capable of assessing instrument performance for peptide separation and analysis in this manner. To create such a standard, we have used the recently described QconCAT methodology to generate an artificial protein, QCAL. This protein, a concatenation of tryptic peptides that is expressed in E. coli, provides a stoichiometrically controlled mixture of peptides that are amenable to analysis by all commonly used instrumentation platforms for proteomics.
Sigma-Aldrich
MS QCAL Peptide Mix, lyophilized powder